Dofetilide 98%

Supplier: Ambeed
Danger

Synonyms: 1-(4-Methanesulphonamidophenoxy)-2-[N-(4-methanesulphonamidophenethyl)-N-methylamino]ethane, N-(4-(2-(Methyl(2-(4-(methylsulphonamido)phenoxy)ethyl)amino)ethyl)phenyl)methanesulphonamide

A253881-10MG A253881-1MG A253881-25MG A253881-50MG A253881-5MG A253881-1G
77153-268EA 10.3 USD
77153-268 77153-270 77153-272 77153-274 77153-276 77685-720
Dofetilide 98%
Dofetilide

Precaución: for Research Use Only

Formula: C₁₉H₂₇N₃O₅S₂
MW: 441.56 g/mol
MDL Number: MFCD00869707
CAS Number: 115256-11-6
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR