4-(Isopropylamino)butanol 96%
Supplier: Ambeed
|
Danger
|
A251273-10G
A251273-1G
A251273-25G
A251273-5G
77153-254EA
50.47
USD
77153-254
77153-256
77153-258
77153-260
4-(Isopropylamino)butanol 96%
4-(Isopropylamino)butanol
Precaución: for Research Use Only
Formula:
C₇H₁₇NO MW: 131.22 g/mol |
MDL Number:
MFCD14708173 CAS Number: 42042-71-7 UN: 3259 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...