21-[(tert-Butoxycarbonyl)amino]-4,7,10,13,16,19-hexaoxaheneicosanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-21-amino-4,7,10,13,16,19-hexaoxaheneicosanoic acid, 2,2-Dimethyl-4-oxo-3,8,11,14,17,20,23-heptaoxa-5-azahexacosan-26-oic acid, Boc-NH-PEG6-CH2CH2COOH, 21-(Boc-amino)-4,7,10,13,16,19-hexaoxaheneicosanoic acid

A250196-5G A250196-1G A250196-100MG A250196-250MG A250196-25G
77153-250EA 299 USD
77153-250 77550-588 77685-709 77901-425 77901-614
21-[(tert-Butoxycarbonyl)amino]-4,7,10,13,16,19-hexaoxaheneicosanoic acid 95%
21-[(tert-Butoxycarbonyl)amino]-4,7,10,13,16,19-hexaoxaheneicosanoic acid

Precaución: for Research Use Only

Formula: C₂₀H₃₉NO₁₀
MDL Number: MFCD07781255
CAS Number: 882847-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR