DL-α-Tocopherol acetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate
A249984-1000G
A249984-100G
A249984-500G
A249984-5G
A249984-25G
77153-242EA
145
USD
77153-242
77153-244
77153-246
77153-248
77685-706
DL-α-Tocopherol acetate 98%
DL-α-Tocopherol acetate
Precaución: for Research Use Only
Formula:
C₃₁H₅₂O₃ MW: 472.75 g/mol |
MDL Number:
MFCD00072042 CAS Number: 7695-91-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...