1-(4-Nitrophenyl)guanidine nitrate 97%

Supplier: Ambeed
Danger

A248923-100MG A248923-1G A248923-250MG A248923-5G
77153-232EA 7.21 USD
77153-232 77153-234 77153-236 77153-238
1-(4-Nitrophenyl)guanidine nitrate 97%
1-(4-Nitrophenyl)guanidine nitrate

Precaución: for Research Use Only

Formula: C₇H₉N₅O₅
MW: 243.18 g/mol
MDL Number: MFCD25958968
CAS Number: 142992-72-1
UN: 1479
ADR: 5.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR