4-Bromo-1-chloro-2-isopropoxybenzene 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1-chloro-2-(propan-2-yloxy)benzene

A248904-1G A248904-250MG
77153-228EA 43.26 USD
77153-228 77153-230
4-Bromo-1-chloro-2-isopropoxybenzene 98%
4-Bromo-1-chloro-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
MW: 249.53 g/mol
MDL Number: MFCD19637696
CAS Number: 637022-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR