Methylene blue trihydrate 97%

Supplier: Ambeed
Danger

Synonyms: 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride trihydrate, C.I.52015, Methylthioninium chloride trihydrate, Basic blue 9, Swiss Blue, Methylene blue

A247644-10G A247644-25G A247644-5G A247644-100G
77153-214EA 7.21 USD
77153-214 77153-216 77153-218 77685-698
Methylene blue trihydrate 97%
Methylene blue trihydrate

Precaución: for Research Use Only

Formula: C₁₆H₁₈ClN₃S·3H₂O
MW: 373.9 g/mol
MDL Number: MFCD00150008
CAS Number: 7220-79-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR