Ethyl 4-amino-3-bromo-5-nitrobenzoate 95%

Supplier: Ambeed
Warning

A246456-100MG A246456-1G A246456-250MG
77153-196EA 7 USD
77153-196 77153-198 77153-200
Ethyl 4-amino-3-bromo-5-nitrobenzoate 95%
Ethyl 4-amino-3-bromo-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O₄
MDL Number: MFCD00044014
CAS Number: 82760-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR