Diphenyleneiodonium chloride 95%

Supplier: Ambeed
Danger

Synonyms: Diphenyleneiodonium Chloride

A246246-1MG A246246-250MG A246246-100MG
77153-192EA 7 USD
77153-192 77685-692 77901-535
Diphenyleneiodonium chloride 95%
Diphenyleneiodonium chloride

Precaución: for Research Use Only

Formula: C₁₂H₈ClI
MDL Number: MFCD00214165
CAS Number: 4673-26-1
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR