(S)-3-((tert-Butoxycarbonyl)amino)-5-(methylthio)pentanoic acid 97%

Supplier: Ambeed
Warning

A245403-100MG A245403-1G
77153-178EA 5 USD
77153-178 77900-780
(S)-3-((tert-Butoxycarbonyl)amino)-5-(methylthio)pentanoic acid 97%
(S)-3-((tert-Butoxycarbonyl)amino)-5-(methylthio)pentanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄S
MDL Number: MFCD06202399
CAS Number: 1217811-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR