1-(2-Aminoethyl)-4-methylpiperazine 97%

Supplier: Ambeed
Danger

Synonyms: 2-(4-Methylpiperazin-1-yl)ethanamine, 2-(4-Methyl-piperazin-1-yl)-ethylamine

A245391-10G A245391-1G A245391-250MG A245391-5G
77153-170EA 88.58 USD
77153-170 77153-172 77153-174 77153-176
1-(2-Aminoethyl)-4-methylpiperazine 97%
1-(2-Aminoethyl)-4-methylpiperazine

Precaución: for Research Use Only

Formula: C₇H₁₇N₃
MW: 143.23 g/mol
MDL Number: MFCD03701701
CAS Number: 934-98-5
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR