3,5-Diamino-1H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Diamino-1,2,4-triazole, Guanazole

A245318-100G A245318-10G A245318-25G A245318-500G A245318-5G A245318-1000G
77153-160EA 34 USD
77153-160 77153-162 77153-164 77153-166 77153-168 77685-688
3,5-Diamino-1H-1,2,4-triazole 98%
3,5-Diamino-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₂H₅N₅
MW: 99.1 g/mol
MDL Number: MFCD00005233
CAS Number: 1455-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR