4-Cyclopropyl-1H-pyrazole 95%
Supplier: Ambeed
|
Warning
|
A242901-1G
A242901-250MG
A242901-100MG
77153-128EA
322.99
USD
77153-128
77153-130
77892-434
4-Cyclopropyl-1H-pyrazole 95%
4-Cyclopropyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₆H₈N₂ |
MDL Number:
MFCD22681810 CAS Number: 90253-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...