1,3-Dichloro-5-fluorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichlorofluorobenzene

A242391-10G A242391-1G A242391-25G
77153-108EA 23.69 USD
77153-108 77153-110 77153-112
1,3-Dichloro-5-fluorobenzene 98%
1,3-Dichloro-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃Cl₂F
MW: 164.9900 g/mol
MDL Number: MFCD00060652
CAS Number: 1435-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR