MnTBAP chloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
MnTBAP, [5,10,15,20-Tetrakis(4-carboxyphenyl)-21H,23H-porphine]manganese(III) chloride
A241977-100MG
A241977-250MG
77153-100EA
99
USD
77153-100
77153-102
MnTBAP chloride 95%
MnTBAP chloride
Precaución: for Research Use Only
Formula:
C₄₈H₂₈MnN₄O₈Cl MW: 879.2 g/mol |
MDL Number:
MFCD06227760 CAS Number: 55266-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...