Boc-Thr-OBzl 98%

Supplier: Ambeed
Warning

Synonyms: (2S,3R)-Benzyl 2-((tert-butoxycarbonyl)amino)-3-hydroxybutanoate

A241579-100G A241579-10G A241579-1G A241579-25G A241579-5G
77153-080EA 152 USD
77153-080 77153-082 77153-084 77153-086 77153-088
Boc-Thr-OBzl 98%
Boc-Thr-OBzl

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₅
MDL Number: MFCD11974981
CAS Number: 33662-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR