Atglistatin 98+%

Supplier: Ambeed
Warning

Synonyms: 3-(4'-(Dimethylamino)-[1,1'-biphenyl]-3-yl)-1,1-dimethylurea, 3-[3-[4-(dimethylamino)phenyl]phenyl]-1,1-dimethylurea

A241114-10MG A241114-1MG A241114-50MG A241114-5MG
77153-072EA 135.96 USD
77153-072 77153-074 77153-076 77153-078
Atglistatin 98+%
Atglistatin

Precaución: for Research Use Only

Formula: C₁₇H₂₁N₃O
MW: 283.37 g/mol
MDL Number: MFCD28009494
CAS Number: 1469924-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR