3,5-Dihydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-chromen-4-one 95%

Supplier: Ambeed
Warning

A239568-10MG A239568-1MG A239568-5MG
77153-056EA 113 USD
77153-056 77153-058 77153-060
3,5-Dihydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-chromen-4-one 95%
3,5-Dihydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₆
MW: 300.26 g/mol
MDL Number: MFCD03419285
CAS Number: 569-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR