2-Amino-4-bromo-6-fluorobenzonitrile 98%

Supplier: Ambeed
Warning

A238179-1G A238179-250MG A238179-5G A238179-25G A238179-10G A238179-100G
77153-034EA 18 USD
77153-034 77153-036 77153-038 77550-534 77685-663 77892-414
2-Amino-4-bromo-6-fluorobenzonitrile 98%
2-Amino-4-bromo-6-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MDL Number: MFCD28155022
CAS Number: 1279865-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR