(R)-4-(((9H-Fluoren-9-yl)methoxy)carbonyl)morpholine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A237525-1G A237525-250MG
77153-026EA 448.05 USD
77153-026 77153-028
(R)-4-(((9H-Fluoren-9-yl)methoxy)carbonyl)morpholine-3-carboxylic acid 97%
(R)-4-(((9H-Fluoren-9-yl)methoxy)carbonyl)morpholine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₅
MW: 353.36 g/mol
MDL Number: MFCD06809895
CAS Number: 942153-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR