trans-3,4-Difluoropyrrolidine hydrochloride 98%

Supplier: Ambeed
Warning

A237442-100MG A237442-1G A237442-250MG
77153-020EA 46 USD
77153-020 77153-022 77153-024
trans-3,4-Difluoropyrrolidine hydrochloride 98%
trans-3,4-Difluoropyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₈ClF₂N
MDL Number: MFCD27997405
CAS Number: 869481-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR