N-(2-(2-Methyl-1H-indol-3-yl)ethyl)thiophene-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: CK-636

A237397-10MG A237397-1MG A237397-5MG
77153-014EA 18.54 USD
77153-014 77153-016 77153-018
N-(2-(2-Methyl-1H-indol-3-yl)ethyl)thiophene-2-carboxamide 98%
N-(2-(2-Methyl-1H-indol-3-yl)ethyl)thiophene-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂OS
MW: 284.37 g/mol
MDL Number: MFCD03036245
CAS Number: 442632-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR