2,5-Dibromo-4-fluorobenzonitrile 97%

Supplier: Ambeed
Warning

A235845-1G A235845-250MG A235845-5G
77152-992EA 58 USD
77152-992 77152-994 77152-996
2,5-Dibromo-4-fluorobenzonitrile 97%
2,5-Dibromo-4-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂Br₂FN
MW: 278.90 g/mol
MDL Number: MFCD18906828
CAS Number: 1379326-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR