Methyl 2-chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylate 95%

Supplier: Ambeed
Warning

A234939-100MG A234939-250MG
77152-988EA 59.74 USD
77152-988 77152-990
Methyl 2-chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylate 95%
Methyl 2-chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₆ClN₃O₂
MW: 211.60 g/mol
MDL Number: MFCD20923472
CAS Number: 1352396-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR