6-Bromo-1H-indazol-5-ol 97%

Supplier: Ambeed
Warning

A234907-10G A234907-1G A234907-250MG A234907-25G A234907-5G
77152-978EA 275 USD
77152-978 77152-980 77152-982 77152-984 77152-986
6-Bromo-1H-indazol-5-ol 97%
6-Bromo-1H-indazol-5-ol

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
MDL Number: MFCD16877652
CAS Number: 1206800-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR