(S)-Methyl 2-(pyrrolidin-3-yl)acetate hydrochloride 95%

Supplier: Ambeed
Warning

A233397-100MG A233397-250MG A233397-1G
77152-942EA 19 USD
77152-942 77152-944 77900-800
(S)-Methyl 2-(pyrrolidin-3-yl)acetate hydrochloride 95%
(S)-Methyl 2-(pyrrolidin-3-yl)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂
MDL Number: MFCD22377962
CAS Number: 1024038-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR