Ku-60019 99+%

Supplier: Ambeed
Warning

Synonyms: (2R,6S)-rel-2,6-Dimethyl-N-[5-[6-(4-morpholinyl)-4-oxo-4H-pyran-2-yl]-9H-thioxanthen-2-yl]-4-morpholineacetamide, 2-[(2R,6S)-2,6-Dimethyl-4-morpholinyl]-N-{5-[6-(4-morpholinyl)-4-oxo-4H-pyran-2-yl]-9H-thioxanthen-2-yl}acetamide, 4-Morpholineacetamide, 2,6-dimethyl-N-[5-[6-(4-morpholinyl)-4-oxo-4H-pyran-2-yl]-9H-thioxanthen-2-yl]-, (2R,6S), 2-((2R,6S)-2,6-Dimethylmorpholino)-N-(5-(6-morpholino-4-oxo-4H-pyran-2-yl)-9H-thioxanthen-2-yl)acetamide

A231772-10MG A231772-1MG A231772-50MG A231772-5MG
77152-920EA 275.01 USD
77152-920 77152-922 77152-924 77152-926
Ku-60019 99+%
Ku-60019

Precaución: for Research Use Only

Formula: C₃₀H₃₃N₃O₅S
MW: 547.67 g/mol
MDL Number: MFCD18384974
CAS Number: 925701-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR