4-Benzyl-2-morpholinecarboxylic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Benzylmorpholine-2-carboxylic acid hydrochloride

A231409-1G A231409-250MG A231409-25G A231409-5G
77152-912EA 14 USD
77152-912 77152-914 77152-916 77152-918
4-Benzyl-2-morpholinecarboxylic acid hydrochloride 97%
4-Benzyl-2-morpholinecarboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₆ClNO₃
MW: 257.71 g/mol
MDL Number: MFCD02682031
CAS Number: 135072-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR