tert-Butyl 4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 95+%
Supplier: Ambeed
|
Warning
|
A230004-1G
A230004-5G
77152-870EA
35.02
USD
77152-870
77152-872
tert-Butyl 4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 95+%
tert-Butyl 4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₂₆BNO₄ MW: 295.18 g/mol |
MDL Number:
MFCD16628193 CAS Number: 1167991-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...