2,3,4-Trichlorophenylboronic acid 98%

Supplier: Ambeed
Warning

A229440-10G A229440-1G A229440-250MG A229440-25G A229440-5G
77152-860EA 285.31 USD
77152-860 77152-862 77152-864 77152-866 77152-868
2,3,4-Trichlorophenylboronic acid 98%
2,3,4-Trichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BCl₃O₂
MDL Number: MFCD03095112
CAS Number: 352530-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR