2-((Benzyloxy)methyl)propane-1,3-diol 97%

Supplier: Ambeed
Warning

A228186-100MG A228186-1G A228186-250MG A228186-5G
77152-838EA 28.84 USD
77152-838 77152-840 77152-842 77152-844
2-((Benzyloxy)methyl)propane-1,3-diol 97%
2-((Benzyloxy)methyl)propane-1,3-diol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃
MW: 196.24 g/mol
MDL Number: MFCD20482799
CAS Number: 117087-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR