Acid red 52 95%

Supplier: Ambeed
Warning

Synonyms: Food Red No.106, Sodium 4-(3,6-bis(diethylamino)xanthylium-9-yl)benzene-1,3-disulphonate, Kiton Red S, Sulphorhodamine B sodium salt, Lissamine rhodamine B-200

A227373-100G A227373-10G A227373-25G A227373-500G A227373-5G
77152-828EA 32.96 USD
77152-828 77152-830 77152-832 77152-834 77152-836
Acid red 52 95%
Acid red 52

Precaución: for Research Use Only

Formula: C₂₇H₃₀N₂O₇S₂
MW: 580.65 g/mol
MDL Number: MFCD00010180
CAS Number: 3520-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR