4-Bromo-3,5-difluorobenzaldehyde 97%

Supplier: Ambeed
Warning

A226440-100MG A226440-1G A226440-250MG A226440-25G A226440-5G
77152-806EA 23 USD
77152-806 77152-808 77152-810 77152-812 77152-814 77152-816
4-Bromo-3,5-difluorobenzaldehyde 97%
4-Bromo-3,5-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
MDL Number: MFCD16659085
CAS Number: 135564-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR