(1R,4R)-5-Methyl-2,5-diazabicyclo-[2.2.1]heptane dihydrochloride 97%

Supplier: Ambeed
Warning

A226069-1G A226069-250MG A226069-100MG
77152-802EA 116 USD
77152-802 77152-804 77900-676
(1R,4R)-5-Methyl-2,5-diazabicyclo-[2.2.1]heptane dihydrochloride 97%
(1R,4R)-5-Methyl-2,5-diazabicyclo-[2.2.1]heptane dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄Cl₂N₂
MDL Number: MFCD23381624
CAS Number: 1403763-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR