1-(Benzylamino)-2-methylpropan-2-ol 95%

Supplier: Ambeed
Warning

A225945-100MG A225945-1G A225945-250MG A225945-5G
77152-794EA 36.05 USD
77152-794 77152-796 77152-798 77152-800
1-(Benzylamino)-2-methylpropan-2-ol 95%
1-(Benzylamino)-2-methylpropan-2-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO
MW: 179.26 g/mol
MDL Number: MFCD09608008
CAS Number: 80466-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR