N-Fmoc-isoindoline-1-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(((9H-Fluoren-9-yl)methoxy)carbonyl)isoindoline-1-carboxylic acid

A225594-100MG A225594-1G A225594-250MG
77152-778EA 70.04 USD
77152-778 77152-780 77152-782
N-Fmoc-isoindoline-1-carboxylic acid 95%
N-Fmoc-isoindoline-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₄H₁₉NO₄
MW: 385.412 g/mol
MDL Number: MFCD00270204
CAS Number: 204320-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR