7-Bromoisobenzofuran-1(3H)-one 95%

Supplier: Ambeed
Warning

A225167-250MG A225167-1G A225167-100MG A225167-5G
77152-776EA 25 USD
77152-776 77685-613 77901-392 77901-564
7-Bromoisobenzofuran-1(3H)-one 95%
7-Bromoisobenzofuran-1(3H)-one

Precaución: for Research Use Only

Formula: C₈H₅BrO₂
MDL Number: MFCD18204744
CAS Number: 105694-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR