BCTC 99+%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(4-(tert-Butyl)phenyl)-4-(3-chloropyridin-2-yl)piperazine-1-carboxamide
A224797-10MG
A224797-1MG
A224797-25MG
A224797-5MG
77152-766EA
46.35
USD
77152-766
77152-768
77152-770
77152-772
BCTC 99+%
BCTC
Precaución: for Research Use Only
Formula:
C₂₀H₂₅ClN₄O MW: 372.897 g/mol |
MDL Number:
MFCD08690556 CAS Number: 393514-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...