(2S,3S)-Benzyl (1-amino-3-methyl-1-oxopentan-2-yl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: Z-Ile-NH₂

A223991-10G A223991-1G A223991-25G A223991-5G
77152-750EA 28 USD
77152-750 77152-752 77152-754 77152-756
(2S,3S)-Benzyl (1-amino-3-methyl-1-oxopentan-2-yl)carbamate 95%
(2S,3S)-Benzyl (1-amino-3-methyl-1-oxopentan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₃
MW: 264.32 g/mol
MDL Number: MFCD00238471
CAS Number: 86161-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR