2-Methylbenzeneboronic acid-1,3-propanediol ester 98+%

Supplier: Ambeed
Warning

A223776-1G A223776-25G A223776-5G
77152-742EA 20 USD
77152-742 77152-744 77152-746
2-Methylbenzeneboronic acid-1,3-propanediol ester 98+%
2-Methylbenzeneboronic acid-1,3-propanediol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₂
MDL Number: MFCD06659935
CAS Number: 163517-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR