2-Methoxy-5-(trifluoromethyl)benzyl bromide 97%
Supplier: Ambeed
|
Danger
|
A222970-100MG
A222970-1G
77152-732EA
8.24
USD
77152-732
77152-734
2-Methoxy-5-(trifluoromethyl)benzyl bromide 97%
2-Methoxy-5-(trifluoromethyl)benzyl bromide
Precaución: for Research Use Only
Formula:
C₉H₈BrF₃O |
MDL Number:
MFCD06660318 CAS Number: 685126-88-9 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...