TCS JNK 5a 98%

Supplier: Ambeed

Synonyms: N-(3-cyano-4,5,6,7-tetrahydro-1-benzothiophen-2-yl)naphthalene-1-carboxamide, N-(3-Cyano-4,5,6,7-tetrahydrobenzo[b]thiophen-2-yl)-1-naphthamide, c-Jun N-terminal Kinase Inhibitor IX, JNK Inhibitor IX

A222483-10MG A222483-1MG A222483-50MG A222483-5MG
77152-716EA 40 USD
77152-716 77152-718 77152-720 77152-722
TCS JNK 5a 98%
TCS JNK 5a

Precaución: for Research Use Only

Formula: C₂₀H₁₆N₂OS
MW: 332.42 g/mol
MDL Number: MFCD01005305
CAS Number: 312917-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR