Methyl 2-(5-(trifluoromethyl)pyridin-2-yl)acetate 95%

Supplier: Ambeed
Warning

A222325-1G A222325-5G A222325-250MG
77152-712EA 207.03 USD
77152-712 77152-714 77685-597
Methyl 2-(5-(trifluoromethyl)pyridin-2-yl)acetate 95%
Methyl 2-(5-(trifluoromethyl)pyridin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
MDL Number: MFCD28986180
CAS Number: 1810715-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR