(R)-Methyl 2-(pyrrolidin-3-yl)acetate hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A221713-1G
A221713-250MG
77152-700EA
114
USD
77152-700
77152-702
(R)-Methyl 2-(pyrrolidin-3-yl)acetate hydrochloride 98%
(R)-Methyl 2-(pyrrolidin-3-yl)acetate hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₄ClNO₂ |
MDL Number:
MFCD22377963 CAS Number: 1024038-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...