1,3,6,8-Tetrabromocarbazole 95%

Supplier: Ambeed
Warning

A221588-1G A221588-250MG A221588-5G
77152-694EA 43.26 USD
77152-694 77152-696 77152-698
1,3,6,8-Tetrabromocarbazole 95%
1,3,6,8-Tetrabromocarbazole

Precaución: for Research Use Only

Formula: C₁₂H₅Br₄N
MW: 482.80 g/mol
MDL Number: MFCD00218272
CAS Number: 55119-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR