2-Methoxy-4-nitrobenzyl alcohol 97%

Supplier: Ambeed
Warning

A220715-1G A220715-250MG
77152-680EA 45 USD
77152-680 77152-682
2-Methoxy-4-nitrobenzyl alcohol 97%
2-Methoxy-4-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉NO₄
MDL Number: MFCD08277262
CAS Number: 136507-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR