cis-3-Hydroxy-3-methylcyclobutylamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: cis-3-Amino-1-methylcyclobutanol hydrochloride

A219359-100MG A219359-1G A219359-250MG
77152-658EA 31.93 USD
77152-658 77152-660 77152-662
cis-3-Hydroxy-3-methylcyclobutylamine hydrochloride 97%
cis-3-Hydroxy-3-methylcyclobutylamine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO·ClH
MW: 137.608 g/mol
MDL Number: MFCD23105823
CAS Number: 1523606-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR