Methyl 2-chloro-5-methylbenzoate 98%

Supplier: Ambeed
Warning

A218127-1G A218127-5G
77152-648EA 10 USD
77152-648 77152-650
Methyl 2-chloro-5-methylbenzoate 98%
Methyl 2-chloro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MDL Number: MFCD12025214
CAS Number: 16220-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR