2-Bromo-4-fluoro-1-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

A217527-1G A217527-250MG A217527-5G A217527-100MG
77152-638EA 54 USD
77152-638 77152-640 77685-589 77900-777
2-Bromo-4-fluoro-1-(trifluoromethoxy)benzene 98%
2-Bromo-4-fluoro-1-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄O
MDL Number: MFCD11847187
CAS Number: 866633-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR