Iron(II) oxalate dihydrate 99+%

Supplier: Ambeed
Warning

Synonyms: Iron (II) ethanedioate dihydrate, Oxalic acid iron (II) salt dihydrate, Ferrous oxalate dihydrate, Iron(II) oxalate hydrate

A216107-1000G A216107-100G A216107-500G
77152-630EA 60 USD
77152-630 77152-632 77152-634
Iron(II) oxalate dihydrate 99+%
Iron(II) oxalate dihydrate

Precaución: for Research Use Only

Formula: Fe(C₂O₄)·2H₂O
MW: 179.9 g/mol
Melting Pt: 190
MDL Number: MFCD00150040
CAS Number: 6047-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR